• USA Home
  • 175145 - Ethyl perfluorobutyrate

EMAIL THIS PAGE TO A FRIEND
175145 Sigma-Aldrich

Ethyl perfluorobutyrate

97%

Synonym: Ethyl heptafluorobutyrate

  • CAS Number 356-27-4

  • Linear Formula CF3CF2CF2COOCH2CH3

  • Molecular Weight 242.09

  •  Beilstein/REAXYS Number 1794634

  •  EC Number 206-602-1

  •  MDL number MFCD00000434

  •  PubChem Substance ID 24850489

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
Quality Level   100
assay   97%
refractive index   n20/D 1.302 (lit.)
density   1.396 g/mL at 25 °C (lit.)
SMILES string   CCOC(=O)C(F)(F)C(F)(F)C(F)(F)F
InChI   1S/C6H5F7O2/c1-2-15-3(14)4(7,8)5(9,10)6(11,12)13/h2H2,1H3
InChI key   JVHJRIQPDBCRRE-UHFFFAOYSA-N

Description

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3272 3 / PGII
WGK Germany 
WGK 3
RTECS 
ET4200000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?