• USA Home
  • 17624 - 3-(Bromomethyl)-2,4,10-trioxatricyclo[3.3.1.13.7]decane

EMAIL THIS PAGE TO A FRIEND
17624 Sigma-Aldrich

3-(Bromomethyl)-2,4,10-trioxatricyclo[3.3.1.13.7]decane

≥97.0% (HPLC)

Synonym: 3-(Bromomethyl)-2,4,10-trioxaadamantane, Bromo(2,3,10-trioxatricyclo[3.3.1.13.7]dec-3-yl)methane

  • CAS Number 157371-80-7

  • Empirical Formula (Hill Notation) C8H11BrO3

  • Molecular Weight 235.08

  •  MDL number MFCD00077628

  •  PubChem Substance ID 57647654

  •  NACRES NA.22

Purchase

Properties

Related Categories Acetals, Ketals & Ortho Esters, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
InChI Key   CSXFBEUWUQOEEC-KVFPUHGPSA-N
assay   ≥97.0% (HPLC)
mp   95-97 °C
storage temp.   2-8°C

Description

Application

3-(Bromomethyl)-2,4,10-trioxatricyclo[3.3.1.13.7]decane may be used in the chemical synthesis studies.

Packaging

1 g in glass bottle

Other Notes

Masked bromoacetic acid derivative which can be transformed to the stable Grignard or lithium-organic reagents

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?