EMAIL THIS PAGE TO A FRIEND
176257 Sigma-Aldrich

Acetyl fluoride

99%

Purchase

Properties

vapor pressure   15.61 psi ( 20 °C)
assay   99%
bp   20-21 °C (lit.)
mp   −84 °C (lit.)
SMILES string   CC(F)=O

Description

Recommended products

Stainless steel hose adapter Z146838 or stainless steel body mini gas regulator Z513547 is recommended.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGI
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

hose barb

Product #

Description

Add to Cart

Z146838 Hose adapter, stainless steel

regulator

Product #

Description

Add to Cart

Z513547 Aldrich® mini gas regulator, stainless steel body

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?