• USA Home
  • 176419 - Tetrafluoroboric acid diethyl ether complex

EMAIL THIS PAGE TO A FRIEND
176419 Aldrich

Tetrafluoroboric acid diethyl ether complex

85%

Synonym: Fluoroboric acid diethyl ether complex

Purchase

Properties

InChI Key   QLXGKERRYKPCOT-UHFFFAOYSA-N
assay   85%
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3129 8(4.3) / PGII
WGK Germany 
1
Flash Point(F) 
264.2 °F
Flash Point(C) 
129 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?