• USA Home
  • 177075 - 2-Chloro-3-(3-chloro-o-tolyl)propionitrile

EMAIL THIS PAGE TO A FRIEND
177075 Sigma-Aldrich

2-Chloro-3-(3-chloro-o-tolyl)propionitrile

99%

  • CAS Number 21342-85-8

  • Empirical Formula (Hill Notation) C10H9Cl2N

  • Molecular Weight 214.09

  •  MDL number MFCD00001860

Purchase

Properties

InChI Key   YYJUXSGXHHPBTK-UHFFFAOYSA-N
assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?