• USA Home
  • 177245 - trans-2-Dodecenedioic acid

EMAIL THIS PAGE TO A FRIEND
177245 Sigma-Aldrich

trans-2-Dodecenedioic acid

97%

Synonym: 2-Dodecenedioic acid, trans-Traumatic acid, Traumatic acid

  • CAS Number 6402-36-4

  • Linear Formula HO2C(CH2)8CH=CHCO2H

  • Molecular Weight 228.28

  •  Beilstein/REAXYS Number 1725762

  •  EC Number 229-019-4

  •  MDL number MFCD00004443

  •  PubChem Substance ID 24850624

Purchase

Properties

assay   97%
mp   165-167 °C (lit.)

Description

Biochem/physiol Actions

Traumatic acid (1 uM) incubated for 30 minutes with sprouts of 9- to 11-day-old seedlings of pea increases total tyrosine phosphorylation of pea proteins . The effect of traumatic acid has differential effects, with an increase in tyrosine phosphorylation in some proteins, but with a decrease in others.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?