• USA Home
  • 177709 - Praseodymium(III) tris[3-(trifluoromethylhydroxymethylene)-d-camphorate]

EMAIL THIS PAGE TO A FRIEND
177709 Aldrich

Praseodymium(III) tris[3-(trifluoromethylhydroxymethylene)-d-camphorate]

98%

Synonym: Pr(facam)3, Pr(tfc)3, Tris(3-trifluoroacetyl-d-camphorato)praseodym(III), Tris[3-(trifluoromethylhydroxymethylene)-d-camphorato]praseodymium(III)

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Resolution Reagents, Chiral Shift Reagents, NMR - Buffers and Reagents,
InChI Key   FCCKUPHNOGBUBW-KDMOJBFPSA-K
assay   98%
optical activity   [α]25/D +175°, c = 1.3 in chloroform
mp   208-210 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?