EMAIL THIS PAGE TO A FRIEND
177946 Sigma-Aldrich

Daminozide

99%

Synonym: Succinic acid mono(2,2-dimethylhydrazide)

  • CAS Number 1596-84-5

  • Linear Formula HO2CCH2CH2CONHN(CH3)2

  • Molecular Weight 160.17

  •  Beilstein/REAXYS Number 1863230

  •  EC Number 216-485-9

  •  MDL number MFCD00002787

Purchase

Properties

assay   99%
mp   162-164 °C (lit.)
SMILES string   CN(C)NC(=O)CCC(O)=O
InChI   1S/C6H12N2O3/c1-8(2)7-5(9)3-4-6(10)11/h3-4H2,1-2H3,(H,7,9)(H,10,11)
InChI key   NOQGZXFMHARMLW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
WM9625000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?