• USA Home
  • 178705 - 1,1,4,4-Tetraphenyl-1,3-butadiene

EMAIL THIS PAGE TO A FRIEND
178705 Sigma-Aldrich

1,1,4,4-Tetraphenyl-1,3-butadiene

99%

Synonym: TPB

  • CAS Number 1450-63-1

  • Linear Formula (C6H5)2C=CHCH=C(C6H5)2

  • Molecular Weight 358.47

  •  Beilstein/REAXYS Number 1914229

  •  MDL number MFCD00004766

  •  PubChem Substance ID 329751574

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
assay   99%
mp   207-209 °C (lit.)
SMILES string   c1ccc(cc1)\C(=C\C=C(/c2ccccc2)c3ccccc3)c4ccccc4
InChI   1S/C28H22/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)21-22-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26/h1-22H
InChI key   KLCLIOISYBHYDZ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?