• USA Home
  • 17887 - Ethyl (1R,2S)-cis-2-hydroxycyclohexanecarboxylate

EMAIL THIS PAGE TO A FRIEND
17887 Sigma-Aldrich

Ethyl (1R,2S)-cis-2-hydroxycyclohexanecarboxylate

≥97.0% (GC)

  • CAS Number 61586-78-5

  • Empirical Formula (Hill Notation) C9H16O3

  • Molecular Weight 172.22

  •  Beilstein/REAXYS Number 2690619

  •  MDL number MFCD02683037

  •  PubChem Substance ID 24850776

Purchase

Properties

assay   ≥97.0% (GC)
optical activity   [α]20/D +25±2°, c = 1% in chloroform
optical purity   enantiomeric ratio: ≥96.5:3.5 (GC)
refractive index   n20/D 1.463

Description

Other Notes

Chiral building block

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?