• USA Home
  • 181196 - Poly(Bisphenol A-co-epichlorohydrin)

EMAIL THIS PAGE TO A FRIEND
181196 Aldrich

Poly(Bisphenol A-co-epichlorohydrin)

average Mw ~40,000, pellets

Purchase

Properties

Related Categories Bisphenol A Polymers and Epoxy Prepolymer Resins, Engineering Polymers, Materials Science, Polymer Science, Polymers More...
form   pellets
mol wt   average Mw ~40,000
loss   <2.00% loss on drying
density   1.18 g/mL at 25 °C(lit.)

Description

Other Notes

May contain varying amounts of epichlorohydrin

Packaging

250 g in poly bottle

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Same Products - New Packaging
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?