• USA Home
  • 18184 - (R)-tert-Butyl ethyl 3-(tert-butyldimethylsiloxy)glutarate

EMAIL THIS PAGE TO A FRIEND
18184 Sigma-Aldrich

(R)-tert-Butyl ethyl 3-(tert-butyldimethylsiloxy)glutarate

≥98.0% (GC)

Synonym: (R)-tert-Butyl ethyl 3-(tert-butyldimethylsiloxy)pentanedioate

Purchase

Properties

assay   ≥98.0% (GC)
density   0.960 g/mL at 20 °C (lit.)

Description

Other Notes

Preparation of rosuvastatin and related HMG-CoA reductase inhibitors

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?