• USA Home
  • 182222 - Poly(methyl acrylate) solution

EMAIL THIS PAGE TO A FRIEND
182222 Sigma-Aldrich

Poly(methyl acrylate) solution

~40 wt. % (in toluene), average Mw 30,000 (Typical), average Mn 10,000 (Typical), secondary standard

Purchase

Properties

grade   secondary standard
vapor density   >1 (vs air)
assay   ~40 wt. % (in toluene)
autoignition temp.   ~896 °F
mol wt   average Mn 10,000 (Typical)
  average Mw 30,000 (Typical)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1294 - class 3 - PG 2 - Toluene, solution
WGK Germany 
3
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

182214 Poly(methyl acrylate) solution, average Mw ~40,000 by GPC

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?