• USA Home
  • 184985 - 2-Bromo-3-nitronaphthalene-bis(hexachlorocyclopentadiene) adduct

EMAIL THIS PAGE TO A FRIEND
184985

2-Bromo-3-nitronaphthalene-bis(hexachlorocyclopentadiene) adduct

technical grade

  • CAS Number 84605-00-5

  • Empirical Formula (Hill Notation) C20H6BrCl12NO2

  • Molecular Weight 797.61

  •  MDL number MFCD00167985

Purchase

Properties

grade   technical grade
SMILES string   [O-][N+](=O)c1cc2C3C(C4C(c2cc1Br)[C@@]5(Cl)C(Cl)=C(Cl)[C@]4(Cl)C5(Cl)Cl)[C@@]6(Cl)C(Cl)=C(Cl)[C@]3(Cl)C6(Cl)Cl

Description

Application

Precursor to substituted naphthalenes which are otherwise difficult to synthesize.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?