• USA Home
  • 18709 - (S)-Boc-4-(trifluoromethyl)-β-Phe-OH

EMAIL THIS PAGE TO A FRIEND
18709 Sigma-Aldrich

(S)-Boc-4-(trifluoromethyl)-β-Phe-OH

≥98.0% (HPLC)

Synonym: (S)-3-(Boc-amino)-3-[4-(trifluoromethyl)phenyl]propionic acid, Boc-4-(trifluoromethyl)-D-β-phenylalanine

  • CAS Number 500770-79-6

  • Empirical Formula (Hill Notation) C15H18F3NO4

  • Molecular Weight 333.30

  •  MDL number MFCD03427927

  •  eCl@ss 32160406

  •  PubChem Substance ID 57647758

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
assay   ≥98.0% (HPLC)
application(s)   peptide synthesis: suitable
SMILES string   CC(C)(C)OC(=O)N[C@@H](CC(O)=O)c1ccc(cc1)C(F)(F)F
InChI   1S/C15H18F3NO4/c1-14(2,3)23-13(22)19-11(8-12(20)21)9-4-6-10(7-5-9)15(16,17)18/h4-7,11H,8H2,1-3H3,(H,19,22)(H,20,21)/t11-/m0/s1
InChI key   WJMVMQYNFZQHGW-NSHDSACASA-N

Description

Packaging

500 mg in poly tube

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?