• USA Home
  • 187178 - 1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
187178 Sigma-Aldrich

1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride

97%

Synonym: 1-Benzyl-4-hydroxyisonipecotonitrile hydrochloride

Purchase

Properties

assay   97%
mp   175-177 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?