• USA Home
  • 187488 - 3-Methyl-1-adamantaneacetic acid

EMAIL THIS PAGE TO A FRIEND
187488 Aldrich

3-Methyl-1-adamantaneacetic acid

99%

  • CAS Number 14202-13-2

  • Empirical Formula (Hill Notation) C13H20O2

  • Molecular Weight 208.30

  •  MDL number MFCD00074754

Purchase

Properties

assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?