• USA Home
  • 187631 - 3-Methyl-4-nitrobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
187631 Sigma-Aldrich

3-Methyl-4-nitrobenzyl alcohol

98%

  • CAS Number 80866-75-7

  • Linear Formula CH3C6H3(NO2)CH2OH

  • Molecular Weight 167.16

  •  Beilstein/REAXYS Number 2363298

  •  EC Number 279-578-3

  •  MDL number MFCD00007171

  •  PubChem Substance ID 24851281

  •  NACRES NA.22

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
Quality Level   100
assay   98%
mp   57-58 °C (lit.)
SMILES string   Cc1cc(CO)ccc1[N+]([O-])=O
InChI   1S/C8H9NO3/c1-6-4-7(5-10)2-3-8(6)9(11)12/h2-4,10H,5H2,1H3
InChI key   KOVQGYQQVNCUBR-UHFFFAOYSA-N

Description

Application

3-Methyl-4-nitrobenzyl alcohol was used as starting reagent in the synthesis of 3-methyl-4-iodophenylalanine.

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?