• USA Home
  • 188905 - 2-Benzoyloxy-2-phenylmalononitrile

EMAIL THIS PAGE TO A FRIEND
188905

2-Benzoyloxy-2-phenylmalononitrile

98%

Synonym: α,α-Dicyanobenzyl benzoate

  • CAS Number 5467-94-7

  • Empirical Formula (Hill Notation) C16H10N2O2

  • Molecular Weight 262.26

  •  MDL number MFCD00001843

Purchase

Properties

assay   98%
SMILES string   O=C(OC(C#N)(C#N)c1ccccc1)c2ccccc2
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?