• USA Home
  • 188964 - 2-Chloro-5-nitrobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
188964 Sigma-Aldrich

2-Chloro-5-nitrobenzyl alcohol

97%

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
assay   97%
mp   75-79 °C (lit.)
SMILES string   OCc1cc(ccc1Cl)[N+]([O-])=O
InChI   1S/C7H6ClNO3/c8-7-2-1-6(9(11)12)3-5(7)4-10/h1-3,10H,4H2
InChI key   NNLQYDLTFRXAKD-UHFFFAOYSA-N

Description

General description

Density functional theory frequency calculations for 2-chloro-5-nitrobenzyl alcohol has been evaluated by FTIR spectra and FT-Raman spectra.

Application

2-Chloro-5-nitrobenzyl alcohol was used in the preparation of macrobicyclic triphosphazides and tri-λ5-phosphazenes.

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?