EMAIL THIS PAGE TO A FRIEND
18921 Sigma-Aldrich

Butane

≥99.0% (GC)

  • CAS Number 106-97-8

  • Linear Formula CH3CH2CH2CH3

  • Molecular Weight 58.12

  •  Beilstein/REAXYS Number 969129

  •  EC Number 203-448-7

  •  MDL number MFCD00009424

  •  PubChem Substance ID 24851369

Purchase

Properties

InChI Key   IJDNQMDRQITEOD-UHFFFAOYSA-N
assay   ≥99.0% (GC)
impurities   2,2-dimethyl-propane
  isobutane
  ≤50 mg/kg S
bp   −0.5 °C (lit.)
mp   −138 °C (lit.)
density   0.579 g/mL at 20 °C (lit.)

Description

Packaging

pressure tin with 250 mL (net ~145 g)

Other Notes

Sales restrictions may apply

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2037A1 2.2
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?