• USA Home
  • 190381 - 2-Phenylpropyl isobutyrate

EMAIL THIS PAGE TO A FRIEND
190381 Aldrich

2-Phenylpropyl isobutyrate

98%

Synonym: Hydratropyl isobutyrate

Purchase

Properties

InChI Key   LZTCJUAHYXNKRE-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.485 (lit.)
density   0.973 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

W289205 2-Phenylpropyl isobutyrate, ≥97%, FG

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?