• USA Home
  • 190470 - 4-Ethoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
190470 Sigma-Aldrich

4-Ethoxybenzyl alcohol

98%

Purchase

Properties

assay   98%
refractive index   n20/D 1.535 (lit.)
bp   273 °C (lit.)
mp   32-34 °C (lit.)
SMILES string   CCOc1ccc(CO)cc1
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?