• USA Home
  • 191655 - 3,5-Dimethoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
191655 Sigma-Aldrich

3,5-Dimethoxybenzyl alcohol

99%

  • CAS Number 705-76-0

  • Linear Formula (CH3O)2C6H3CH2OH

  • Molecular Weight 168.19

  •  Beilstein/REAXYS Number 1866305

  •  EC Number 211-888-6

  •  MDL number MFCD00004641

  •  PubChem Substance ID 24851502

  •  NACRES NA.22

Purchase

Properties

Related Categories Alcohols, Building Blocks, C9 to C10, Chemical Synthesis, Organic Building Blocks,
assay   99%
mp   43-46 °C (lit.)
SMILES string   COc1cc(CO)cc(OC)c1
InChI   1S/C9H12O3/c1-11-8-3-7(6-10)4-9(5-8)12-2/h3-5,10H,6H2,1-2H3
InChI key   AUDBREYGQOXIFT-UHFFFAOYSA-N

Description

General description

3,5-Dimethoxybenzyl alcohol is an important pharmaceutical intermediate.

Application

3,5-Dimethoxybenzyl alcohol was used as starting material for the synthesis of dendrimeric compounds.

Packaging

5, 25 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?