EMAIL THIS PAGE TO A FRIEND
193194 Sigma-Aldrich

Trimethylene borate

Synonym: 1,3-Propanediol cyclic ester with boric acid

Purchase

Properties

refractive index   n20/D 1.454 (lit.)
bp   125 °C/0.05 mmHg (lit.)
density   1.153 g/mL at 25 °C (lit.)
SMILES string   C(COB1OCCCO1)COB2OCCCO2
InChI   1S/C9H18B2O6/c1-4-12-10(13-5-1)16-8-3-9-17-11-14-6-2-7-15-11/h1-9H2
InChI key   FZNHHPNJQSIEPT-UHFFFAOYSA-N

Description

Application

Reagent for the preparation of alkylboronic acids and esters.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?