• USA Home
  • 194107 - (−)-Dihydrocarvyl acetate

EMAIL THIS PAGE TO A FRIEND
194107 Aldrich

(−)-Dihydrocarvyl acetate

≥95%

  • CAS Number 20777-49-5

  • Empirical Formula (Hill Notation) C12H20O2

  • Molecular Weight 196.29

  •  Beilstein Registry Number 2443836

  •  EC Number 244-029-9

  •  MDL number MFCD00001501

  •  PubChem Substance ID 24851630

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Protected Alcohols More...
InChI Key   TUSIZTVSUSBSQI-UHFFFAOYSA-N
assay   ≥95%
refractive index   n20/D 1.459(lit.)
bp   232-234 °C(lit.)
density   0.947 g/mL at 20 °C(lit.)
Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
OT0210000
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?