EMAIL THIS PAGE TO A FRIEND
195146 Sigma-Aldrich

Mordant Brown 48

Dye content 20 %

Synonym: 2-(5-Chloro-2,4-diaminophenylazo)-4,6-dinitrophenol

  • CAS Number 6232-53-7

  • Linear Formula ClC6H2(NH2)2N=NC6H2(NO2)2OH

  • Molecular Weight 352.69

  •  Colour Index Number 11300

  •  EC Number 228-338-6

  •  MDL number MFCD00007802

  •  PubChem Substance ID 24851674

Purchase

Properties

composition   Dye content, 20%
absorption   λmax 492 nm
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?