• USA Home
  • 195383 - 1-(Trifluoromethyl)ethenyl acetate

EMAIL THIS PAGE TO A FRIEND
195383 Aldrich

1-(Trifluoromethyl)ethenyl acetate

99%

Purchase

Properties

assay   99%
refractive index   n20/D 1.341(lit.)
bp   86 °C(lit.)
density   1.212 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
Flash Point(F) 
37.4 °F
Flash Point(C) 
3 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?