EMAIL THIS PAGE TO A FRIEND
195642 Sigma-Aldrich

7-Ethoxycoumarin

99.5%

Synonym: 7-Ethoxy-1-benzopyran-2-one

  • CAS Number 31005-02-4

  • Empirical Formula (Hill Notation) C11H10O3

  • Molecular Weight 190.20

  •  Beilstein/REAXYS Number 146200

  •  EC Number 250-429-4

  •  MDL number MFCD00006877

  •  PubChem Substance ID 329751917

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Coumarins,
assay   99.5%
mp   88-90 °C (lit.)
SMILES string   CCOc1ccc2C=CC(=O)Oc2c1
InChI   1S/C11H10O3/c1-2-13-9-5-3-8-4-6-11(12)14-10(8)7-9/h3-7H,2H2,1H3
InChI key   LIFAQMGORKPVDH-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

100 mg in glass bottle

Application

Excellent substrate for a rapid, sensitive, fluorometric assay of microsomal monooxygenase activity.

Biochem/physiol Actions

Substrate for CYP2B6

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

E1379 7-Ethoxycoumarin

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?