• USA Home
  • 195693 - 2,2,2-Tribromoethyl phosphoromorpholinochloridate

EMAIL THIS PAGE TO A FRIEND
195693 Sigma-Aldrich

2,2,2-Tribromoethyl phosphoromorpholinochloridate

Purchase

Properties

mp   80-83 °C (lit.)
SMILES string   ClP(=O)(OCC(Br)(Br)Br)N1CCOCC1
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?