• USA Home
  • 195766 - Benzoin isopropyl ether

EMAIL THIS PAGE TO A FRIEND
195766 Sigma-Aldrich

Benzoin isopropyl ether

98%

Synonym: α-Isopropoxy-α-phenylacetophenone

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   MSAHTMIQULFMRG-UHFFFAOYSA-N
assay   98%
bp   175-180 °C/10 mmHg(lit.)
mp   78-80 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?