• USA Home
  • 199354 - 1-[4-(Dimethylamino)phenyl]-2-thiourea

EMAIL THIS PAGE TO A FRIEND
199354 Aldrich

1-[4-(Dimethylamino)phenyl]-2-thiourea

98%

  • CAS Number 22283-43-8

  • Empirical Formula (Hill Notation) C9H13N3S

  • Molecular Weight 195.28

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?