EMAIL THIS PAGE TO A FRIEND
19939 Sigma-Aldrich

N-tert-Butylurea

≥97.0%

  • CAS Number 1118-12-3

  • Linear Formula (CH3)3CNHCONH2

  • Molecular Weight 116.16

  •  Beilstein/REAXYS Number 1744501

  •  EC Number 214-257-3

  •  MDL number MFCD00025448

  •  PubChem Substance ID 57647811

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks, Ureas More...
assay   ≥97.0%
mp   ~185 °C (dec.)
SMILES string   CC(C)(C)NC(N)=O
InChI   1S/C5H12N2O/c1-5(2,3)7-4(6)8/h1-3H3,(H3,6,7,8)
InChI key   JLEHSYHLHLHPAL-UHFFFAOYSA-N

Description

Application

N-tert-Butylurea was used in the synthesis of substituted ureas.

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
YS3680000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?