• USA Home
  • 19971 - Phenylbis[4-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)phenyl]phosphine

EMAIL THIS PAGE TO A FRIEND
19971 Sigma-Aldrich

Phenylbis[4-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)phenyl]phosphine

≥90.0%

Synonym: Bis[4-(1H,1H,2H,2H-perfluorooctyl)phenyl]phenylphosphine

  • CAS Number 290827-94-0

  • Empirical Formula (Hill Notation) C34H21F26P

  • Molecular Weight 954.46

  •  MDL number MFCD03788364

  •  PubChem Substance ID 57647812

  •  NACRES NA.22

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Fluorous Synthesis, Phosphine Compounds, Phosphine Ligands,
assay   ≥90.0%
mp   46-51 °C
functional group   phosphine
reaction suitability   reaction type: Buchwald-Hartwig Cross Coupling Reaction
  reaction type: Heck Reaction
  reaction type: Hiyama Coupling
  reaction type: Negishi Coupling
  reaction type: Sonogashira Coupling
  reaction type: Stille Coupling
  reaction type: Suzuki-Miyaura Coupling
  reagent type: ligand
reaction type: Fluorous synthesis
SMILES string   FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCc1ccc(cc1)P(c2ccccc2)c3ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc3
InChI   1S/C34H21F26P/c35-23(36,25(39,40)27(43,44)29(47,48)31(51,52)33(55,56)57)16-14-18-6-10-21(11-7-18)61(20-4-2-1-3-5-20)22-12-8-19(9-13-22)15-17-24(37,38)26(41,42)28(45,46)30(49,50)32(53,54)34(58,59)60/h1-13H,14-17H2
InChI key   GNCTUKGIBHXUPH-UHFFFAOYSA-N

Description

Packaging

1, 5 g in poly bottle

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?