EMAIL THIS PAGE TO A FRIEND
199745 Sigma-Aldrich

Ponceau SS

Dye content 80 %

Synonym: Acid Red 150

  • CAS Number 6226-78-4

  • Empirical Formula (Hill Notation) C22H14N4Na2O7S2

  • Molecular Weight 556.48

  •  Colour Index Number 27190

  •  EC Number 228-318-7

  •  MDL number MFCD00003906

  •  PubChem Substance ID 24851936

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, P, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   YDGHROMBRLEXLZ-UHFFFAOYSA-L
composition   Dye content, 80%
absorption   λmax 351 nm (2nd)
  λmax 514 nm

Description

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?