• USA Home
  • 200263 - Hydroxybutyl methyl cellulose

EMAIL THIS PAGE TO A FRIEND
200263 Sigma-Aldrich

Hydroxybutyl methyl cellulose

average Mn ~120,000, powder

  • CAS Number 9041-56-9

  • Empirical Formula (Hill Notation) C64H124O30

  • Molecular Weight 1373.65

Purchase

Properties

form   powder
mol wt   average Mn ~120,000
extent of labeling   3.5% hydroxybutyl
viscosity   14,000 cP(lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?