• USA Home
  • 201006 - N-(Chlorocarbonyl)isocyanate

EMAIL THIS PAGE TO A FRIEND
201006 Sigma-Aldrich

N-(Chlorocarbonyl)isocyanate

technical grade

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks More...
grade   technical grade
InChI Key   VBBUFMFZDHLELS-UHFFFAOYSA-N
refractive index   n20/D 1.432 (lit.)
bp   63 °C(lit.)
density   1.31 g/mL at 25 °C (lit.)

Description

Application

For a review of the reactions of this versatile synthon.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?