EMAIL THIS PAGE TO A FRIEND
201081 Sigma-Aldrich

Quadricyclane

99%

Synonym: Quadricyclo[2.2.1.02,6.03,5]heptane

Purchase

Properties

assay   99%
SMILES string   C1[C@H]2[C@@H]3[C@H]2[C@@H]4[C@H]1[C@H]34
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3384 3(6.1) / PGI
WGK Germany 
3
RTECS 
XB7436000
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?