• USA Home
  • 201693 - Phenolphthalein carbinol disulfate tripotassium salt hydrate

EMAIL THIS PAGE TO A FRIEND
201693 Aldrich

Phenolphthalein carbinol disulfate tripotassium salt hydrate

Purchase

Properties

InChI Key   ANNXSRCLBOSUSB-UHFFFAOYSA-K
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?