EMAIL THIS PAGE TO A FRIEND
201839 Aldrich

Acid Blue 161

Dye content 40 %

  • CAS Number 12392-64-2

  • Linear Formula C20H13N2O5SNaCrx

  • Molecular Weight 394.40

  •  Colour Index Number 15706

  •  MDL number MFCD00151053

Purchase

Properties

composition   Dye content, 40%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?