• USA Home
  • 203777 - Mercury(II) chloride

EMAIL THIS PAGE TO A FRIEND
203777 Aldrich

Mercury(II) chloride

99.999% trace metals basis

Synonym: Mercuric chloride

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Hg, Materials Science,
vapor pressure   1.3 mmHg ( 236 °C)
InChI Key   LWJROJCJINYWOX-UHFFFAOYSA-L
assay   99.999% trace metals basis
form   solid
mp   277 °C (lit.)

Description

Packaging

50 g in poly bottle

Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?