• USA Home
  • 204145 - Praseodymium(III,IV) oxide

EMAIL THIS PAGE TO A FRIEND
204145 Aldrich

Praseodymium(III,IV) oxide

99.999% trace metals basis

Purchase

Properties

Related Categories Alternative Energy, Catalysis and Inorganic Chemistry, Chemical Synthesis, Dopants, Materials Science,
InChI Key   AICMLAQRDYRMRP-UHFFFAOYSA-N
assay   99.999% trace metals basis
form   powder
density   6.5 g/mL at 25 °C(lit.)

Description

Features and Benefits

Used in the solid state synthesis of a remarkable ternary, reduced molybdenum oxide, Pr4Mo9O18, whose structure contains previously unknown Mo7, Mo13and Mo19 clusters. The new cluster product is a small band gap semiconductor.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TU1480000
Protocols & Articles

Articles

Metal Hydrides for NiMH Battery Applications

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications. The Partnership for a New Generation of V...
Dhanesh Chandra,* Wen-Ming Chien and Anjali Talekar
Material Matters Volume 6 Article 2
Keywords: Absorption, Automotive, Capillary electrophoresis, Combustion, Degradations, Electrochemical analysis, Electronics, Environmental, Ion-exchange chromatography, Phase transitions, Precipitation, Purification, Separation, Substitutions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?