• USA Home
  • 204587 - Terbium(III) sulfate octahydrate

EMAIL THIS PAGE TO A FRIEND
204587 Sigma-Aldrich

Terbium(III) sulfate octahydrate

99.999%

Purchase

Properties

InChI Key   SRSFZSUHOIHUFR-UHFFFAOYSA-H
assay   99.999%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

325953 Terbium(III) sulfate octahydrate, 99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?