• USA Home
  • 205362 - Dimethyl nitroterephthalate

EMAIL THIS PAGE TO A FRIEND
205362 Sigma-Aldrich

Dimethyl nitroterephthalate

99%

Synonym: Dimethyl 2-nitrobenzene-1,4-dicarboxylate

  • CAS Number 5292-45-5

  • Linear Formula O2NC6H3-1,4-(CO2CH3)2

  • Molecular Weight 239.18

  •  Beilstein/REAXYS Number 1993064

  •  EC Number 226-134-1

  •  MDL number MFCD00008422

  •  PubChem Substance ID 24852327

Purchase

Properties

assay   99%
mp   73-76 °C (lit.)
SMILES string   COC(=O)c1ccc(C(=O)OC)c(c1)[N+]([O-])=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?