• USA Home
  • 20550 - (S)-3-tert-Butyl-2,5-piperazinedione

EMAIL THIS PAGE TO A FRIEND
20550 Sigma-Aldrich

(S)-3-tert-Butyl-2,5-piperazinedione

≥99.0% (CHN)

  • CAS Number 65050-07-9

  • Empirical Formula (Hill Notation) C8H14N2O2

  • Molecular Weight 170.21

  •  Beilstein/REAXYS Number 4396135

  •  MDL number MFCD00077675

  •  PubChem Substance ID 57647845

  •  NACRES NA.22

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks, Piperazines More...
assay   ≥99.0% (CHN)
optical activity   [α]20/D +78±1°, c = 1% in acetic acid
storage temp.   2-8°C
SMILES string   CC(C)(C)[C@@H]1NC(=O)CNC1=O
InChI   1S/C8H14N2O2/c1-8(2,3)6-7(12)9-4-5(11)10-6/h6H,4H2,1-3H3,(H,9,12)(H,10,11)/t6-/m1/s1
InChI key   OWQNUQNZSDAFHT-ZCFIWIBFSA-N

Description

Packaging

1 g in glass bottle

Other Notes

Derivative of L-tert-leucine used in the bislactim ether method for the preparation of amino acids in very high enantioselectivity

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?