• USA Home
  • 209910 - 2-Methoxytetrahydrofuran

EMAIL THIS PAGE TO A FRIEND
209910 Aldrich

2-Methoxytetrahydrofuran

99%

  • CAS Number 13436-45-8

  • Empirical Formula (Hill Notation) C5H10O2

  • Molecular Weight 102.13

  •  PubChem Substance ID 329752068

Purchase

Properties

InChI Key   OKAMTPRCXVGTND-UHFFFAOYSA-N
assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?