EMAIL THIS PAGE TO A FRIEND
210277 Aldrich

D-allo-Threonine

99%

Synonym: (2R,3R)-2-Amino-3-hydroxybutyric acid

  • CAS Number 24830-94-2

  • Linear Formula CH3CH(OH)CH(NH2)CO2H

  • Molecular Weight 119.12

  •  Beilstein/REAXYS Number 1721644

  •  EC Number 246-488-0

  •  MDL number MFCD00004526

Purchase

Properties

assay   99%
optical activity   [α]23/D −8.8°, c = 2 in H2O
mp   276 °C (dec.) (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
BA4050000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?