• USA Home
  • 211346 - (S)-(+)-1-(9-Anthryl)-2,2,2-trifluoroethanol

EMAIL THIS PAGE TO A FRIEND
211346 Sigma-Aldrich

(S)-(+)-1-(9-Anthryl)-2,2,2-trifluoroethanol

≥98%

Synonym: (S)-(+)-α-(Trifluoromethyl)anthracene-9-methanol, (S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol

  • CAS Number 60646-30-2

  • Empirical Formula (Hill Notation) C16H11F3O

  • Molecular Weight 276.25

  •  Beilstein/REAXYS Number 4695162

  •  EC Number 262-348-1

  •  MDL number MFCD00062968

  •  PubChem Substance ID 24852670

  •  NACRES NA.22

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   ICZHJFWIOPYQCA-HNNXBMFYSA-N
assay   ≥98%
optical activity   [α]25/D +29°, c = 6.3 in chloroform
mp   132-135 °C (lit.)

Description

General description

(S)-(+)-1-(9-Anthryl)-2,2,2-trifluoroethanol is a chiral solvating agent (CSA).

Application

(S)-(+)-1-(9-Anthryl)-2,2,2-trifluoroethanol has been used for the NMR spectral determination of optical purity (and in some cases, absolute configuration) of a wide variety of sulfoxides, lactones, amines, sulfinate esters, oxaziridines and allenes.

Packaging

100 mg in clear glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?