• USA Home
  • 212822 - Lithium hydroxide monohydrate

EMAIL THIS PAGE TO A FRIEND
212822 Sigma-Aldrich

Lithium hydroxide monohydrate

reagent grade, 98%

Purchase

Properties

grade   reagent grade
assay   98%
SMILES string   [Li+].O.[OH-]
InChI   1S/Li.2H2O/h;2*1H2/q+1;;/p-1
InChI key   GLXDVVHUTZTUQK-UHFFFAOYSA-M

Description

General description

Crystals of Lithium hydroxide monohydrate belong to the monoclinic crystal system. It can be prepared by reacting Li metal with H2O, followed by the evaporation of the aqueous solution.

Application

Lithium hydroxide monohydrate (LiOH.H2O) may be employed as a catalyst for the melt transesterification of diphenyl carbonate and bisphenol A. It can also be used for the synthesis of 3,4-disubstituted pyrrole derivatives.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2680 8 / PGII
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?