• USA Home
  • 213985 - 2-Methoxyphenylacetone

EMAIL THIS PAGE TO A FRIEND
213985 Sigma-Aldrich

2-Methoxyphenylacetone

98%

Purchase

Properties

assay   98%
refractive index   n20/D 1.525 (lit.)
bp   127-130 °C/10 mmHg(lit.)
density   1.054 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?